Assay Details
Gene Symbol: LOC5576030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655921.2 | XP_001655971.2 | 2 - 3 | 308 | 69 | |
XM_021846922.1 | XP_021702614.1 | 2 - 3 | 258 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001655921.2, XM_021846922.1 transcripts.
Set Membership: |
![]() ![]() |
