Assay Details
Gene Symbol: LOC5576030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655921.2 | XP_001655971.2 | 3 - 4 | 4450 | 78 | |
XM_021846922.1 | XP_021702614.1 | 3 - 4 | 4400 | 78 |
