Assay Details
Gene Symbol: LOC5573640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660916.3 | XP_001660966.3 | 1 - 2 | 789 | 72 | |
XM_021837700.1 | XP_021693392.1 | 1 - 2 | 789 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001660916.3, XM_021837700.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
