Assay Details
Gene Symbol: LOC5574099 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840407.1 | XP_021696099.1 | 4 - 5 | 1619 | 59 | |
XM_021840408.1 | XP_021696100.1 | 5 - 6 | 1526 | 59 | ||
XM_021840409.1 | XP_021696101.1 | 5 - 6 | 1315 | 59 | ||
XM_021840410.1 | XP_021696102.1 | 5 - 6 | 1130 | 59 | ||
XM_021840411.1 | XP_021696103.1 | 5 - 6 | 1248 | 59 | ||
XM_021840412.1 | XP_021696104.1 | 6 - 7 | 1651 | 59 | ||
XM_021840413.1 | XP_021696105.1 | 6 - 7 | 1488 | 59 | ||
XM_021840414.1 | XP_021696106.1 | 4 - 5 | 974 | 59 | ||
XM_021840415.1 | XP_021696107.1 | 5 - 6 | 1156 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
