Assay Details
Gene Symbol: LOC5566349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845081.1 | XP_021700773.1 | 1 - 2 | 1174 | 85 | |
XM_021845082.1 | XP_021700774.1 | 2 - 3 | 963 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845081.1, XM_021845082.1 transcripts.
Set Membership: |
![]() ![]() |
