Assay Details
Gene Symbol: LOC5566349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845081.1 | XP_021700773.1 | 2 - 3 | 1351 | 64 | |
XM_021845082.1 | XP_021700774.1 | 3 - 4 | 1140 | 64 |

More Information
Set Membership: |
![]() ![]() |
