Assay Details
Gene Symbol: LOC5566349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845081.1 | XP_021700773.1 | 3 - 4 | 1698 | 90 | |
XM_021845082.1 | XP_021700774.1 | 4 - 5 | 1487 | 90 | ||
XM_021845083.1 | XP_021700775.1 | 2 - 3 | 453 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845083.1 transcripts.
Set Membership: |
![]() ![]() |
