Assay Details
Gene Symbol: LOC5566349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845081.1 | XP_021700773.1 | 4 - 5 | 1997 | 81 | |
XM_021845082.1 | XP_021700774.1 | 5 - 6 | 1786 | 81 | ||
XM_021845083.1 | XP_021700775.1 | 3 - 4 | 752 | 81 | ||
XM_021845084.1 | XP_021700776.1 | 2 - 3 | 421 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
