Assay Details
Gene Symbol: LOC5564855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649124.2 | XP_001649174.2 | 5 - 6 | 1144 | 84 | |
XM_021845252.1 | XP_021700944.1 | 6 - 7 | 1356 | 84 | ||
XM_021845253.1 | XP_021700945.1 | 5 - 6 | 1128 | 84 | ||
XM_021845254.1 | XP_021700946.1 | 5 - 6 | 1120 | 84 | ||
XM_021845255.1 | XP_021700947.1 | 5 - 6 | 1397 | 84 | ||
XM_021845256.1 | XP_021700948.1 | 5 - 6 | 1020 | 84 | ||
XM_021845257.1 | XP_021700949.1 | 5 - 6 | 1143 | 84 | ||
XM_021845259.1 | XP_021700951.1 | 5 - 6 | 1142 | 84 | ||
XM_021845260.1 | XP_021700952.1 | 5 - 6 | 1143 | 84 | ||
XM_021845261.1 | XP_021700953.1 | 5 - 6 | 1144 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
