Assay Details
Gene Symbol: LOC5564855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649124.2 | XP_001649174.2 | 4 - 5 | 854 | 77 | |
XM_021845252.1 | XP_021700944.1 | 5 - 6 | 1066 | 77 | ||
XM_021845253.1 | XP_021700945.1 | 4 - 5 | 838 | 77 | ||
XM_021845254.1 | XP_021700946.1 | 4 - 5 | 830 | 77 | ||
XM_021845255.1 | XP_021700947.1 | 4 - 5 | 1107 | 77 | ||
XM_021845256.1 | XP_021700948.1 | 4 - 5 | 730 | 77 | ||
XM_021845257.1 | XP_021700949.1 | 4 - 5 | 853 | 77 | ||
XM_021845259.1 | XP_021700951.1 | 4 - 5 | 852 | 77 | ||
XM_021845260.1 | XP_021700952.1 | 4 - 5 | 853 | 77 | ||
XM_021845261.1 | XP_021700953.1 | 4 - 5 | 854 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
