Assay Details
Gene Symbol: LOC5564855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649124.2 | XP_001649174.2 | 2 - 3 | 283 | 106 | |
XM_021845252.1 | XP_021700944.1 | 3 - 4 | 495 | 106 | ||
XM_021845253.1 | XP_021700945.1 | 2 - 3 | 267 | 106 | ||
XM_021845254.1 | XP_021700946.1 | 2 - 3 | 259 | 106 | ||
XM_021845255.1 | XP_021700947.1 | 2 - 3 | 536 | 106 | ||
XM_021845256.1 | XP_021700948.1 | 2 - 3 | 159 | 106 | ||
XM_021845257.1 | XP_021700949.1 | 2 - 3 | 282 | 106 | ||
XM_021845259.1 | XP_021700951.1 | 2 - 3 | 281 | 106 | ||
XM_021845260.1 | XP_021700952.1 | 2 - 3 | 282 | 106 | ||
XM_021845261.1 | XP_021700953.1 | 2 - 3 | 283 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001649124.2, XM_021845252.1, XM_021845253.1, XM_021845254.1, XM_021845255.1, XM_021845256.1, XM_021845257.1, XM_021845259.1, XM_021845260.1, XM_021845261.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
