Assay Details
Gene Symbol: LOC5564855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649124.2 | XP_001649174.2 | 1 - 2 | 147 | 85 | |
XM_021845257.1 | XP_021700949.1 | 1 - 2 | 146 | 85 | ||
XM_021845259.1 | XP_021700951.1 | 1 - 2 | 145 | 85 | ||
XM_021845260.1 | XP_021700952.1 | 1 - 2 | 146 | 85 | ||
XM_021845261.1 | XP_021700953.1 | 1 - 2 | 147 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001649124.2, XM_021845257.1, XM_021845259.1, XM_021845260.1, XM_021845261.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
