Assay Details
Gene Symbol: LOC5564855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649124.2 | XP_001649174.2 | 10 - 11 | 2496 | 76 | |
XM_021845252.1 | XP_021700944.1 | 11 - 12 | 2708 | 76 | ||
XM_021845253.1 | XP_021700945.1 | 10 - 11 | 2480 | 76 | ||
XM_021845254.1 | XP_021700946.1 | 10 - 11 | 2472 | 76 | ||
XM_021845255.1 | XP_021700947.1 | 10 - 11 | 2749 | 76 | ||
XM_021845256.1 | XP_021700948.1 | 10 - 11 | 2372 | 76 | ||
XM_021845257.1 | XP_021700949.1 | 10 - 11 | 2495 | 76 | ||
XM_021845259.1 | XP_021700951.1 | 10 - 11 | 2494 | 76 | ||
XM_021845260.1 | XP_021700952.1 | 10 - 11 | 2495 | 76 | ||
XM_021845261.1 | XP_021700953.1 | 10 - 11 | 2496 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
