Assay Details
Gene Symbol: LOC5572154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660178.2 | XP_001660228.2 | 3 - 4 | 699 | 60 | |
XM_001660179.2 | XP_001660229.2 | 3 - 4 | 706 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001660178.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
