Assay Details
Gene Symbol: LOC5570958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659376.2 | XP_001659426.1 | 5 - 6 | 1196 | 78 | |
XM_021845805.1 | XP_021701497.1 | 6 - 7 | 1340 | 78 | ||
GenBank mRNA | KJ598054.1 | - | 5 - 6 | 1201 | 78 | |
KJ598055.1 | - | 6 - 7 | 1345 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
