Assay Details
Gene Symbol: LOC5570958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659376.2 | XP_001659426.1 | 3 - 4 | 640 | 79 | |
XM_021845805.1 | XP_021701497.1 | 3 - 4 | 640 | 79 | ||
XM_021845806.1 | XP_021701498.1 | 3 - 4 | 624 | 79 | ||
GenBank mRNA | KJ598054.1 | - | 3 - 4 | 645 | 79 | |
KJ598055.1 | - | 3 - 4 | 645 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
