Assay Details
Gene Symbol: LOC5577549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847098.1 | XP_021702790.1 | 4 - 5 | 2326 | 57 | |
XM_021847099.1 | XP_021702791.1 | 5 - 6 | 2432 | 57 | ||
XM_021847100.1 | XP_021702792.1 | 5 - 6 | 2375 | 57 | ||
XM_021847101.1 | XP_021702793.1 | 4 - 5 | 2231 | 57 | ||
XM_021847103.1 | XP_021702795.1 | 4 - 5 | 2149 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
