Assay Details
Gene Symbol: LOC5564179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648449.2 | XP_001648499.1 | 1 - 2 | 789 | 77 | |
XM_021853567.1 | XP_021709259.1 | 2 - 3 | 587 | 77 | ||
XM_021853568.1 | XP_021709260.1 | 2 - 3 | 557 | 77 | ||
XM_021853569.1 | XP_021709261.1 | 2 - 3 | 311 | 77 | ||
XM_021853570.1 | XP_021709262.1 | 1 - 2 | 793 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001648449.2, XM_021853567.1, XM_021853568.1, XM_021853569.1, XM_021853570.1 transcripts.
Set Membership: |
![]() ![]() |
