Assay Details
Gene Symbol: LOC5564179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648449.2 | XP_001648499.1 | 4 - 5 | 1078 | 76 | |
XM_021853567.1 | XP_021709259.1 | 5 - 6 | 876 | 76 | ||
XM_021853568.1 | XP_021709260.1 | 5 - 6 | 846 | 76 | ||
XM_021853569.1 | XP_021709261.1 | 5 - 6 | 600 | 76 |
