Assay Details
Gene Symbol: LOC5574468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655078.3 | XP_001655128.2 | 4 - 5 | 1975 | 74 | |
XM_021843923.1 | XP_021699615.1 | 5 - 6 | 1325 | 74 | ||
XM_021843924.1 | XP_021699616.1 | 5 - 6 | 1262 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
