Assay Details
Gene Symbol: LOC5569240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857577.1 | XP_021713269.1 | 12 - 13 | 4308 | 81 | |
XM_021857578.1 | XP_021713270.1 | 12 - 13 | 4269 | 81 | ||
XM_021857580.1 | XP_021713272.1 | 12 - 13 | 3960 | 81 | ||
XM_021857581.1 | XP_021713273.1 | 12 - 13 | 3789 | 81 | ||
XM_021857582.1 | XP_021713274.1 | 12 - 13 | 3864 | 81 |
