Assay Details
Gene Symbol: LOC5569240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857577.1 | XP_021713269.1 | 18 - 19 | 5384 | 91 | |
XM_021857578.1 | XP_021713270.1 | 18 - 19 | 5345 | 91 | ||
XM_021857580.1 | XP_021713272.1 | 18 - 19 | 5036 | 91 | ||
XM_021857581.1 | XP_021713273.1 | 18 - 19 | 4865 | 91 | ||
XM_021857582.1 | XP_021713274.1 | 18 - 19 | 4940 | 91 |
