Assay Details
Gene Symbol: LOC5569240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857577.1 | XP_021713269.1 | 20 - 21 | 5917 | 89 | |
XM_021857578.1 | XP_021713270.1 | 20 - 21 | 5878 | 89 | ||
XM_021857580.1 | XP_021713272.1 | 20 - 21 | 5569 | 89 | ||
XM_021857581.1 | XP_021713273.1 | 20 - 21 | 5398 | 89 | ||
XM_021857582.1 | XP_021713274.1 | 20 - 21 | 5473 | 89 |
