Assay Details
Gene Symbol: LOC5569240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857577.1 | XP_021713269.1 | 2 - 3 | 904 | 89 | |
XM_021857578.1 | XP_021713270.1 | 2 - 3 | 865 | 89 | ||
XM_021857580.1 | XP_021713272.1 | 2 - 3 | 556 | 89 | ||
XM_021857581.1 | XP_021713273.1 | 2 - 3 | 385 | 89 | ||
XM_021857582.1 | XP_021713274.1 | 2 - 3 | 460 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021857577.1, XM_021857578.1, XM_021857580.1, XM_021857581.1, XM_021857582.1 transcripts.
Set Membership: |
![]() ![]() |
