Assay Details
Gene Symbol: LOC5569240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857577.1 | XP_021713269.1 | 8 - 9 | 2924 | 65 | |
XM_021857578.1 | XP_021713270.1 | 8 - 9 | 2885 | 65 | ||
XM_021857580.1 | XP_021713272.1 | 8 - 9 | 2576 | 65 | ||
XM_021857581.1 | XP_021713273.1 | 8 - 9 | 2405 | 65 | ||
XM_021857582.1 | XP_021713274.1 | 8 - 9 | 2480 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
