Assay Details
Gene Symbol: LOC5573450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847959.1 | XP_021703651.1 | 3 - 4 | 2991 | 86 | |
XM_021847960.1 | XP_021703652.1 | 3 - 4 | 2991 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
