Assay Details
Gene Symbol: LOC5573299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843374.1 | XP_021699066.1 | 2 - 3 | 789 | 67 | |
XM_021843375.1 | XP_021699067.1 | 2 - 3 | 885 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
