Assay Details
Gene Symbol: LOC5573853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845596.1 | XP_021701288.1 | 1 - 2 | 206 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845596.1 transcripts.
Set Membership: |
![]() ![]() |
