Assay Details
Gene Symbol: LOC5566365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650703.2 | XP_001650753.2 | 1 - 2 | 87 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001650703.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
