Assay Details
Gene Symbol: LOC5566365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650703.2 | XP_001650753.2 | 3 - 4 | 1841 | 94 | |
XM_021850453.1 | XP_021706145.1 | 4 - 5 | 2025 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
