Assay Details
Gene Symbol: LOC110679692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855265.1 | XP_021710957.1 | 6 - 7 | 2112 | 61 | |
XM_021855266.1 | XP_021710958.1 | 6 - 7 | 2074 | 61 | ||
XM_021855267.1 | XP_021710959.1 | 6 - 7 | 1987 | 61 | ||
XM_021855268.1 | XP_021710960.1 | 6 - 7 | 1942 | 61 | ||
XM_021855269.1 | XP_021710961.1 | 6 - 7 | 1583 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
