Assay Details
Gene Symbol: LOC5569008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850328.1 | XP_021706020.1 | 6 - 7 | 2272 | 62 | |
XM_021850329.1 | XP_021706021.1 | 6 - 7 | 2686 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
