Assay Details
Gene Symbol: LOC5572946 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851797.1 | XP_021707489.1 | 1 - 2 | 582 | 58 | |
XM_021851805.1 | XP_021707497.1 | 1 - 2 | 582 | 58 | ||
XM_021851806.1 | XP_021707498.1 | 1 - 2 | 582 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021851797.1, XM_021851805.1, XM_021851806.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
