Assay Details
Gene Symbol: LOC5572946 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851797.1 | XP_021707489.1 | 5 - 6 | 4376 | 65 | |
XM_021851798.1 | XP_021707490.1 | 5 - 6 | 4349 | 65 | ||
XM_021851799.1 | XP_021707491.1 | 5 - 6 | 4125 | 65 | ||
XM_021851800.1 | XP_021707492.1 | 5 - 6 | 4219 | 65 | ||
XM_021851801.1 | XP_021707493.1 | 5 - 6 | 4223 | 65 | ||
XM_021851802.1 | XP_021707494.1 | 5 - 6 | 3890 | 65 | ||
XM_021851804.1 | XP_021707496.1 | 5 - 6 | 3968 | 65 | ||
XM_021851806.1 | XP_021707498.1 | 5 - 6 | 4376 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
