Assay Details
Gene Symbol: LOC5572946 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851797.1 | XP_021707489.1 | 6 - 7 | 4676 | 106 | |
XM_021851798.1 | XP_021707490.1 | 6 - 7 | 4649 | 106 | ||
XM_021851799.1 | XP_021707491.1 | 6 - 7 | 4425 | 106 | ||
XM_021851800.1 | XP_021707492.1 | 6 - 7 | 4519 | 106 | ||
XM_021851801.1 | XP_021707493.1 | 6 - 7 | 4523 | 106 | ||
XM_021851802.1 | XP_021707494.1 | 6 - 7 | 4190 | 106 | ||
XM_021851804.1 | XP_021707496.1 | 6 - 7 | 4268 | 106 | ||
XM_021851805.1 | XP_021707497.1 | 6 - 7 | 4655 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
