Assay Details
Gene Symbol: LOC5564988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649272.3 | XP_001649322.2 | 4 - 5 | 1390 | 80 | |
XM_021838271.1 | XP_021693963.1 | 4 - 5 | 1387 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
