Assay Details
Gene Symbol: LOC5564988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649272.3 | XP_001649322.2 | 3 - 4 | 1206 | 74 | |
XM_021838271.1 | XP_021693963.1 | 3 - 4 | 1203 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
