Assay Details
Gene Symbol: LOC5571886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843131.1 | XP_021698823.1 | 2 - 3 | 1008 | 106 | |
XM_021843132.1 | XP_021698824.1 | 2 - 3 | 1008 | 106 |

More Information
Set Membership: |
![]() ![]() |
