Assay Details
Gene Symbol: LOC23687408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838806.1 | XP_021694498.1 | 2 - 3 | 716 | 69 | |
XM_021838807.1 | XP_021694499.1 | 2 - 3 | 699 | 69 | ||
XM_021838808.1 | XP_021694500.1 | 2 - 3 | 602 | 69 | ||
XM_021838809.1 | XP_021694501.1 | 2 - 3 | 617 | 69 | ||
XM_021838810.1 | XP_021694502.1 | 2 - 3 | 716 | 69 | ||
XM_021838811.1 | XP_021694503.1 | 2 - 3 | 715 | 69 | ||
XM_021838812.1 | XP_021694504.1 | 2 - 3 | 715 | 69 | ||
XM_021838813.1 | XP_021694505.1 | 2 - 3 | 715 | 69 | ||
XM_021838814.1 | XP_021694506.1 | 2 - 3 | 715 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021838806.1, XM_021838807.1, XM_021838808.1, XM_021838809.1, XM_021838810.1, XM_021838811.1, XM_021838812.1, XM_021838813.1, XM_021838814.1 transcripts.
Set Membership: |
![]() ![]() |
