Assay Details
Gene Symbol: LOC23687408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838806.1 | XP_021694498.1 | 3 - 4 | 836 | 79 | |
XM_021838807.1 | XP_021694499.1 | 3 - 4 | 819 | 79 | ||
XM_021838808.1 | XP_021694500.1 | 3 - 4 | 722 | 79 | ||
XM_021838809.1 | XP_021694501.1 | 3 - 4 | 737 | 79 | ||
XM_021838810.1 | XP_021694502.1 | 3 - 4 | 836 | 79 | ||
XM_021838811.1 | XP_021694503.1 | 3 - 4 | 835 | 79 | ||
XM_021838812.1 | XP_021694504.1 | 3 - 4 | 835 | 79 | ||
XM_021838813.1 | XP_021694505.1 | 3 - 4 | 835 | 79 | ||
XM_021838814.1 | XP_021694506.1 | 3 - 4 | 835 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
