Assay Details
Gene Symbol: LOC5580226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852541.1 | XP_021708233.1 | 18 - 19 | 3604 | 69 | |
XM_021852543.1 | XP_021708235.1 | 19 - 20 | 3622 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
