Assay Details
Gene Symbol: LOC5580226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852541.1 | XP_021708233.1 | 33 - 34 | 8984 | 71 | |
XM_021852543.1 | XP_021708235.1 | 34 - 35 | 9002 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
