Assay Details
Gene Symbol: LOC5580226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852541.1 | XP_021708233.1 | 31 - 32 | 7658 | 66 | |
XM_021852543.1 | XP_021708235.1 | 32 - 33 | 7676 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
