Assay Details
Gene Symbol: LOC5565782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650080.2 | XP_001650130.2 | 2 - 3 | 323 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001650080.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
