Assay Details
Gene Symbol: LOC5576370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839737.1 | XP_021695429.1 | 1 - 2 | 236 | 77 | |
XM_021839743.1 | XP_021695435.1 | 1 - 2 | 236 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021839737.1, XM_021839743.1 transcripts.
Set Membership: |
![]() ![]() |
