Assay Details
Gene Symbol: LOC5576370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839723.1 | XP_021695415.1 | 6 - 7 | 2131 | 82 | |
XM_021839733.1 | XP_021695425.1 | 6 - 7 | 2132 | 82 | ||
XM_021839737.1 | XP_021695429.1 | 2 - 3 | 734 | 82 | ||
XM_021839743.1 | XP_021695435.1 | 2 - 3 | 734 | 82 |
