Assay Details
Gene Symbol: LOC5576370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839723.1 | XP_021695415.1 | 7 - 8 | 2353 | 95 | |
XM_021839737.1 | XP_021695429.1 | 3 - 4 | 956 | 95 |
