Assay Details
Gene Symbol: LOC5576370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839723.1 | XP_021695415.1 | 8 - 9 | 2438 | 91 | |
XM_021839737.1 | XP_021695429.1 | 4 - 5 | 1041 | 91 |
