Assay Details
Gene Symbol: LOC5569176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845337.1 | XP_021701029.1 | 3 - 4 | 1968 | 67 | |
XM_021845338.1 | XP_021701030.1 | 3 - 4 | 1803 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845337.1, XM_021845338.1 transcripts.
Set Membership: |
![]() ![]() |
