Assay Details
Gene Symbol: LOC5569176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845337.1 | XP_021701029.1 | 17 - 18 | 4522 | 64 | |
XM_021845338.1 | XP_021701030.1 | 17 - 18 | 4357 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
